(S)-(+)-Ethyl-3-hydroxybutyrate

Warning

Synonyms: Ethyl (S)-(+)-3-hydroxybutyrateEthyl (S)-3-hydroxybutyrate(S)-Ethyl-3-hydroxybutyrate(S)-Ethyl 3-hydroxybutanoate

J63184-06 J63184-03 J63184-03 J63184-06
AAJ63184-06EA 378 USD
CAAAJ63184-03 CAAAJ63184-06 AAJ63184-06 AAJ63184-03
(S)-(+)-Ethyl-3-hydroxybutyrate
(S)-(+)-Ethyl 3-hydroxybutyrate

For synthesis of optically active products


Liquid

Formula: C₆H₁₂O₃
MW: 132.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066206
CAS Number: 56816-01-4
EINECS: 260-393-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR