Famotidine ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: 3-(((2-Guanidinothiazol-4-yl)methyl)thio)-N-sulfamoylpropanimidamide3-(((2-Guanidinothiazol-4-yl)methyl)thio)-N-sulphamoylpropanimidamide3-(2-Guanidinothiazol-4-ylmethylthio)-N1-sulfamoylpropionamide3-(2-Guanidinothiazol-4-ylmethylthio)-N1-sulphamoylpropionamide

J63165-03 J63165-14 J63165-06 J63165-14 J63165-03 J63165-06
AAJ63165-06EA 255.63 USD
AAJ63165-06 CAAAJ63165-14 CAAAJ63165-03 AAJ63165-03 AAJ63165-14 CAAAJ63165-06
Famotidine ≥98%
Famotidine

A competitive histamine H2 receptor antagonist


Powder

Formula: C₈H₁₅N₇O₂S₃
MW: 337.44 g/mol
Melting Pt: 166…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00079297
CAS Number: 76824-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR