Ebastine

Supplier: Thermo Scientific Chemicals

Synonyms: 4-(4-(Benzhydryloxy)piperidin-1-yl)-1-(4-(tert-butyl)phenyl)butan-1-one

J63088-06 J63088-03 J63088-03 J63088-06
AAJ63088-03EA 170.45 USD
CAAAJ63088-03 AAJ63088-03 CAAAJ63088-06 AAJ63088-06
Ebastine
Ebastine

A histamine H1 receptor antagonist


Powder

Formula: C₃₂H₃₉NO₂
MW: 469.67 g/mol
Melting Pt: 84…87 °C
Storage Temperature: Ambient
MDL Number: MFCD00865661
CAS Number: 90729-43-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR