HEPES, 0.5M buffer soln., pH 8.0

Supplier: Alfa Aesar
J63002-AK J63002-AE J63002-AK J63002-AE
AAJ63002-AKEA 168.27 USD
AAJ63002-AK CAAAJ63002-AE CAAAJ63002-AK AAJ63002-AE
HEPES, 0.5M buffer soln., pH 8.0
Buffers Biological Buffers
Liquid

CAS No.: 7365-45-9
Physical Form: Liquid
MDL No.: MFCD00006158
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR