GBR 12935 dihydrochloride

J62991-MA J62991-MA J62991-MF J62991-MF
AAJ62991-MAEA 112.59 USD
AAJ62991-MA CAAAJ62991-MA CAAAJ62991-MF AAJ62991-MF
GBR 12935 dihydrochloride
GBR 12935 dihydrochloride

A very potent and selective inhibitor of dopamine uptake


Powder

Formula: C₂₈H₃₄N₂O·2HCl
MW: 487.51 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00083175
CAS Number: 67469-81-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR