Tricine, 0.5M buffer soln., pH 8.0

Supplier: Alfa Aesar
J62983-AK J62983-AE J62983-AE J62983-AK
AAJ62983-AKEA 105.93 USD
AAJ62983-AK CAAAJ62983-AE AAJ62983-AE CAAAJ62983-AK
Tricine, 0.5M buffer soln., pH 8.0
Buffers Biological Buffers
Liquid

CAS No.: 5704-04-1
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00004277
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR