Ethyl orange sodium salt
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Diethylaminoazobenzene-4'-sulfonic Acid Sodium Salt
J62863-22
J62863-36
J62863-36
J62863-22
AAJ62863-36EA
160.01
USD
AAJ62863-36
AAJ62863-22
CAAAJ62863-22
CAAAJ62863-36
Ethyl orange sodium salt
Ethyl orange sodium salt
Indicator: pH 3.0 (red) to pH 4.5 (yellow)
Powder
Formula:
C₁₆H₁₈N₃NaO₃S MW: 355.39 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00007503 CAS Number: 62758-12-7 EINECS: 263-716-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...