Trimyristin ≥95%

Supplier: Alfa Aesar

Synonyms: Glycerol Trimyristate

J62819-14 J62819-14
AAJ62819-14EA 298.03 USD InStock
CAAAJ62819-14 AAJ62819-14
Trimyristin ≥95%
Trimyristin

Powder

Formula: C₄₅H₈₆O₆
MW: 723.18 g/mol
Melting Pt: 56…57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00036229
CAS Number: 555-45-3
EINECS: 209-099-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR