TRIS, 1.0M buffer soln., pH 8.0

Supplier: Alfa Aesar
J62726-K2 J62726-K2
AAJ62726-K2EA 68.68 USD InStock
CAAAJ62726-K2 AAJ62726-K2
TRIS, 1.0M buffer soln., pH 8.0
Buffers Biological Buffers
Liquid

CAS No.: 77-86-1
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00004679
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR