MES, 0.5M buffer soln., pH 6.0

Supplier: Alfa Aesar
J62574-AK J62574-AE J62574-AE J62574-AK
AAJ62574-AKEA 101.39 USD InStock
AAJ62574-AK CAAAJ62574-AE AAJ62574-AE CAAAJ62574-AK
MES, 0.5M buffer soln., pH 6.0
Buffers Biological Buffers
Liquid

CAS No.: 145224-94-8
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00283848
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR