TRIS, 1.0M buffer soln., pH 6.5, 0.2 micron filtered

Supplier: Alfa Aesar
J62264-AP J62264-AP
AAJ62264-APEA 100.13 USD
CAAAJ62264-AP AAJ62264-AP
TRIS, 1.0M buffer soln., pH 6.5, 0.2 micron filtered
Buffers Biological Buffers
Liquid

CAS No.: 77-86-1
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00004679
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR