Potassium phosphate, 0.2M buffer soln., pH 7.5

Supplier: Alfa Aesar
J62239-K2 J62239-K2 J62239-AP J62239-AP
AAJ62239-K2EA 152.89 USD
CAAAJ62239-K2 AAJ62239-K2 AAJ62239-AP CAAAJ62239-AP
Potassium phosphate, 0.2M buffer soln., pH 7.5
Buffers Biological Buffers
Liquid

CAS No.: 7778-77-0
Physical Form: Liquid
MDL No.: MFCD00011401
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR