Blocking Reagents, BLOTTO in TBS, with 0.02% Sodium Azide, Alfa Aesar
Supplier: Alfa Aesar
J62235-AK J62235-AP J62235-AP J62235-AK
AAJ62235-APEA 93.32 USD
CAAAJ62235-AK CAAAJ62235-AP AAJ62235-AP AAJ62235-AK
Blocking Reagents, BLOTTO in TBS, with 0.02% Sodium Azide, Alfa Aesar
Buffers Electrophoresis Buffers Blocking Buffers
Blocking reagent in immunohistochemistry experiments.
Contains 5% non-fat dry milk and 0.02% sodium azide in TBS.