Polyethylene glycol-lithium acetate solution
Supplier: Thermo Scientific Chemicals
J62213-AK
J62213-AK
AAJ62213-AKEA
66.24
USD
CAAAJ62213-AK
AAJ62213-AK
Polyethylene glycol-lithium acetate solution
Polyethylene glycol-lithium acetate
Common solution for protein and DNA isolation from yeast
Contains 40% PEG 3350 in solution with 100mM lithium acetate, 10mM Tris-HCl, and 1mM EDTA, pH8.0.