TRIS, 1.0M buffer soln., pH 9.5

Supplier: Alfa Aesar
J62084-K2 J62084-K2
AAJ62084-K2EA 99.07 USD
CAAAJ62084-K2 AAJ62084-K2
TRIS, 1.0M buffer soln., pH 9.5
Buffers Biological Buffers
Liquid

CAS No.: 77-86-1
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00004679
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR