Oligomycin
Supplier: Thermo Scientific Chemicals
Synonyms:
Oligomycin complex
, Oligomycin A, B, C
J61898-MA
J61898-MA
AAJ61898-MAEA
273.85
USD
CAAAJ61898-MA
AAJ61898-MA
Oligomycin
Oligomycin
Inhibits respiration in mitochondria. Also inhibits ATP synthase
Powder
Formula:
C₄₅H₇₄O₁₁ MW: 789.3 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD01779388 CAS Number: 1404-19-9 EINECS: 215-767-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...