MOPS, 1.0M buffer soln., pH 7.0

Supplier: Alfa Aesar
J61821-AP J61821-AK J61821-AP J61821-AK
AAJ61821-APEA 308.55 USD
AAJ61821-AP AAJ61821-AK CAAAJ61821-AP CAAAJ61821-AK
MOPS, 1.0M buffer soln., pH 7.0
Buffers Biological Buffers
Liquid

CAS No.: 1132-61-2
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00006183
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR