PIPES, 0.5M buffer soln., pH 6.8

Supplier: Alfa Aesar
J61786-AE J61786-AK J61786-AK J61786-AE
AAJ61786-AEEA 126.65 USD
AAJ61786-AE AAJ61786-AK CAAAJ61786-AK CAAAJ61786-AE
PIPES, 0.5M buffer soln., pH 6.8
Buffers Biological Buffers
Liquid

CAS No.: 5625-37-6
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00006159
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR