Urease

Supplier: Alfa Aesar
J61455-03 J61455-09 J61455-MD J61455-03 J61455-MD
AAJ61455-09EA 1158.2 USD
CAAAJ61455-03 AAJ61455-09 AAJ61455-MD AAJ61455-03 CAAAJ61455-MD
Urease
Enzymes
MW = 480 kDa

Applications: Catalyzes hydrolysis of urea.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR