TRIS, 1.0M buffer soln., pH 8.0, 0.2 micron filtered

Supplier: Alfa Aesar
J61429-AP J61429-AP
AAJ61429-APEA 85.95 USD InStock
AAJ61429-AP CAAAJ61429-AP
TRIS, 1.0M buffer soln., pH 8.0, 0.2 micron filtered
Buffers Biological Buffers
Liquid

CAS No.: 77-86-1
Physical Form: Liquid
Odor: Odorless
MDL No.: MFCD00004679
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR