Would you like to log in again?
Due to system maintenance, custom pricing and product availability will not be shown from 7:00pm EST Friday through 6:00pm Sunday. Orders placed will be processed after maintenance is complete.
Leu-Enkephalin
Supplier: Alfa Aesar
J61408-MB J61408-MB
AAJ61408-MBEA 95.37 USD InStock
CAAAJ61408-MB AAJ61408-MB
Leu-Enkephalin
Proteins and Peptides
Lyophilized powder
Endogenous opioid neurotransmit
Applications: Endogenous opioid neurotransmitter that is an agonist at mu- and delta-opioid receptors
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty