PIPES, 0.5M buffer soln., pH 8.0

Supplier: Alfa Aesar
J61406-AE J61406-AK J61406-AK J61406-AE
AAJ61406-AKEA 149.03 USD
CAAAJ61406-AE AAJ61406-AK CAAAJ61406-AK AAJ61406-AE
PIPES, 0.5M buffer soln., pH 8.0
Buffers Biological Buffers
Liquid

CAS No.: 5625-37-6
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
MDL No.: MFCD00006159
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR