Citrate, 0.5M buffer soln., pH 3.0

Supplier: Alfa Aesar
J61391-AP J61391-AK J61391-AK J61391-AP
AAJ61391-APEA 147.91 USD
AAJ61391-AP CAAAJ61391-AK AAJ61391-AK CAAAJ61391-AP
Citrate, 0.5M buffer soln., pH 3.0
Buffers Biological Buffers
Liquid

CAS No.: 6132-04-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00150031
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR