EPPS, 0.2M buffer soln., pH 8.0

Supplier: Alfa Aesar
J61296-AE J61296-AK J61296-AE J61296-AK
AAJ61296-AKEA 149.99 USD
CAAAJ61296-AE AAJ61296-AK AAJ61296-AE CAAAJ61296-AK
EPPS, 0.2M buffer soln., pH 8.0
Buffers Biological Buffers
Liquid

CAS No.: 16052-06-5
Physical Form: Liquid
MDL No.: MFCD00006160
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR