TRIS, 0.5M buffer soln., pH 7.5

Supplier: Alfa Aesar
J61141-K2 J61141-AP J61141-AP J61141-K2
AAJ61141-K2EA 124.96 USD
AAJ61141-K2 AAJ61141-AP CAAAJ61141-AP CAAAJ61141-K2
TRIS, 0.5M buffer soln., pH 7.5
Buffers Biological Buffers
Liquid

CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00004679
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR