Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Supplier: Thermo Scientific
J61106-MD J61106-MC J61106-MD
AAJ61106-MDEA 255.8 USD
CAAAJ61106-MD AAJ61106-MD 10814-246
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Proteins and Peptides
Aminopeptidase B inhibitor. Also inhibits leucine aminopeptidase.

  • GHS symbol and signal word: GHS07 and warning
  • Melting point: 233 to 236 °C
  • Merck index: 149,842
  • EINECS: 261-529-2
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR