Acetate, 1M buffer soln., pH, 5.5

Supplier: Alfa Aesar
J61033-AK J61033-AP J61033-AP J61033-AK
AAJ61033-AKEA 99.98 USD
AAJ61033-AK AAJ61033-AP CAAAJ61033-AP CAAAJ61033-AK
Acetate, 1M buffer soln., pH, 5.5
Buffers Biological Buffers
Liquid

CAS No.: 127-09-3
Physical Form: Liquid
MDL No.: MFCD00012459
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR