Pectin (from citrus)

Supplier: Thermo Scientific Chemicals

J61021-22 J61021-36 J61021-22 J61021-36
AAJ61021-22EA 52.56 USD
CAAAJ61021-22 AAJ61021-22 AAJ61021-36 CAAAJ61021-36
Pectin (from citrus)
Pectin

Powder

Storage Temperature: Ambient
MDL Number: MFCD00081838
CAS Number: 9000-69-5
EINECS: 232-553-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR