Purine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 1H-Purine

J60999-03 J60999-03
AAJ60999-03EA 230.3 USD
AAJ60999-03 CAAAJ60999-03
Purine 98%
Purine

Crystals

Formula: C₅H₄N₄
MW: 120.11 g/mol
Boiling Pt: 265 °C (1013 hPa)
Melting Pt: 215…217 °C
Density: 1.22 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00079221
CAS Number: 120-73-0
EINECS: 204-421-2
Merck Index: 12,08129

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR