3,3'-Diaminobenzidine, tablets

Supplier: Alfa Aesar
Danger

Synonyms: DAB3,3',4,4'-BiphenyltetraminBiphenyl-3,3',4,4'-tetrayltetraamine

J60972-KW J60972-KX J60972-KX J60972-KW
AAJ60972-KXEA 330.57 USD
CAAAJ60972-KW AAJ60972-KX CAAAJ60972-KX AAJ60972-KW
3,3'-Diaminobenzidine, tablets
3,3'-Diaminobenzidine

Tablets


Widely used peroxidase substrate for immunoblotting and immunohistochemical staining techniques. Commonly used for detection of horseradish peroxidase in membrane-bound and in situ applications. Insoluble end product is resistant to alcohol and fading, making it ideal for immunohistochemistry.

Formula: C₁₂H₁₄N₄
MW: 214.27 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00007725
CAS Number: 91-95-2
EINECS: 202-110-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR