NZYM medium

Supplier: Thermo Scientific
J60918-AP J60918-AP J60918-K2 J60918-K2
AAJ60918-APEA 94.01 USD
CAAAJ60918-AP AAJ60918-AP CAAAJ60918-K2 AAJ60918-K2
NZYM medium
Cell Culture Media
Liquid

Physical Form: Liquid

Applications: NZYM Medium is used for cultivating recombinant strains of Escherichia coli
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR