HEPES, 1.0M buffer soln., pH 7.5

Supplier: Alfa Aesar
J60712-AP J60712-AK J60712-AK J60712-AP
AAJ60712-APEA 252.38 USD
AAJ60712-AP CAAAJ60712-AK AAJ60712-AK CAAAJ60712-AP
HEPES, 1.0M buffer soln., pH 7.5
Buffers Biological Buffers
Liquid

CAS No.: 7365-45-9
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
MDL No.: MFCD00006158
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR