1,3-Dihydroxyacetone dimer

Supplier: Thermo Scientific Chemicals

Synonyms: 2,5-Dihydroxy-1,4-dioxane-2,5-dimethanol

J60592-06 J60592-06
AAJ60592-06EA 28.91 USD
CAAAJ60592-06 AAJ60592-06
1,3-Dihydroxyacetone dimer
1,3-Dihydroxyacetone dimer

Powder

Formula: C₃H₆O₃
MW: 90.08 g/mol
Melting Pt: 75…80 °C
Density: 1,52 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00051019
CAS Number: 96-26-4
EINECS: 202-494-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR