Pyronin G 0.2% (w/v) in aqueous solution

Warning

Synonyms: 3,6-Bis(dimethylamino)xanthylium chloride Pyronine Y

J60505-AB J60505-AB J60505-AC J60505-AC
AAJ60505-ACEA 59.61 USD
CAAAJ60505-AC CAAAJ60505-AB AAJ60505-AC AAJ60505-AB
Pyronin G 0.2% (w/v) in aqueous solution
Pyronine G

Liquid

Formula: C₁₇H₁₉ClN₂O
MW: 302.8 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011725
CAS Number: 92-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR