Acetate, 1M buffer soln., pH, 3.5

Supplier: Alfa Aesar
J60340-AK J60340-AP J60340-AP J60340-AK
AAJ60340-APEA 159.54 USD
CAAAJ60340-AK AAJ60340-AP CAAAJ60340-AP AAJ60340-AK
Acetate, 1M buffer soln., pH, 3.5
Buffers Biological Buffers
Liquid

CAS No.: 127-09-3
Physical Form: Liquid
MDL No.: MFCD00012459
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR