Agarose gel loading dye 6X for electrophoresis, sucrose based
Supplier: Alfa Aesar
J60333-AD J60333-AC J60333-AD J60333-AC
AAJ60333-ACEA 66.58 USD
CAAAJ60333-AD AAJ60333-AC AAJ60333-AD CAAAJ60333-AC
Agarose gel loading dye 6X for electrophoresis, sucrose based
Stains Electrophoresis Stains
Contains 0.25% bromophenol blue, 0.25% xylene cyanol FF, 40% sSucrose (w/v).