STE buffer soln.

Supplier: Alfa Aesar
J60265-K2 J60265-K3 J60265-K2 J60265-K3
AAJ60265-K2EA 47.63 USD
CAAAJ60265-K2 CAAAJ60265-K3 AAJ60265-K2 AAJ60265-K3
STE buffer soln.
Buffers Extraction Buffers
STE buffer is a DNA resuspension and storage buffer used in plasmid Kits for plasmid purification and in some plasmid supplementary protocols.

  • Used in DNA extraction, DNA electrophoresis and many other lab protocols
  • Soluble in water
  • Contains 100mM NaCl, 10mM Tris-HCl, 1mM EDTA, pH 8.0

Physical Form: Liquid.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR