Citrate, 0.5M buffer soln., pH 5.0

Supplier: Alfa Aesar
J60125-AP J60125-AP J60125-AK J60125-AK
AAJ60125-APEA 151.89 USD
CAAAJ60125-AP AAJ60125-AP AAJ60125-AK CAAAJ60125-AK
Citrate, 0.5M buffer soln., pH 5.0
Buffers Biological Buffers
Liquid

CAS No.: 6132-04-3
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Colorless
Odor: Odorless
MDL No.: MFCD00150031
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR