TRIS-buffered saline (TBS, 10X, low salt) pH 8.0

Supplier: Alfa Aesar
J60102-K3 J60102-K3 J60102-K2 J60102-K2 J60102-K7 J60102-K7
AAJ60102-K3EA 63.85 USD InStock
CAAAJ60102-K3 AAJ60102-K3 CAAAJ60102-K2 AAJ60102-K2 CAAAJ60102-K7 AAJ60102-K7
TRIS-buffered saline (TBS, 10X, low salt) pH 8.0
Buffers Electrophoresis Buffers Washing Buffers
Liquid

CAS No.: 77-86-1
Physical Form: Liquid
MDL No.: MFCD00004679
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR