Transformation Reagent, LiSorb, Thermo Scientific

Supplier: Thermo Scientific
LiSorb
J60087-AP J60087-AK J60087-AP J60087-AK
AAJ60087-APEA 57.34 USD
AAJ60087-AP CAAAJ60087-AP CAAAJ60087-AK AAJ60087-AK
Transformation Reagent, LiSorb, Thermo Scientific
Nucleic Acid Reagents Transformation Reagents
Used for transformation of competent cells.

  • Liquid in form
  • Store at ambient

Contains 100 mM lithium acetate, 50 mM Tris-HCl, 1 mM EDTA and 1 mM sorbitol, pH 8.0.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR