3-(2-Methylphenoxy)piperidine hydrochloride 98%

Warning

H59396-MD H59396-03
AAH59396-03EA 215.34 USD
AAH59396-03 AAH59396-MD
3-(2-Methylphenoxy)piperidine hydrochloride 98%
3-(2-Methylphenoxy)piperidine hydrochloride
Formula: C₁₂H₁₈ClNO
Storage Temperature: Ambient
MDL Number: MFCD21606599
CAS Number: 1858256-18-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR