3-Chloro-N,N-dimethylbenzamide 97%

Warning

H58964-ME H58964-MC
AAH58964-MCEA 44.96 USD
AAH58964-MC AAH58964-ME
3-Chloro-N,N-dimethylbenzamide 97%
3-Chloro-N,N-dimethylbenzamide
Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01214035
CAS Number: 24167-52-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR