2-(4-Methylphenyl)benzimidazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(p-Tolyl)-1H-benzo[d]imidazole

H58894-04 H58894-09
AAH58894-09EA 659.34 USD
AAH58894-09 AAH58894-04
2-(4-Methylphenyl)benzimidazole 97%
2-(4-Methylphenyl)benzimidazole
Formula: C₁₄H₁₂N₂
MW: 208.26 g/mol
Melting Pt: 265…269 °C
Storage Temperature: Ambient
MDL Number: MFCD00209485
CAS Number: 120-03-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR