1,6-Bis(maleimido)hexane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N,N'-Hexamethylenebismaleimide1,1'-(Hexane-1,6-diyl)bis(1H-pyrrole-2,5-dione)1,1'-(hexane-1,6-diyl)bis(1h-pyrrole-2,5-dione)N,N'-Hexamethylenedimaleimide1,6-Di(maleimido)hexaneBismaleimidohexaneBMH

H58874-ME H58874-MC
AAH58874-MCEA 151.99 USD
AAH58874-MC AAH58874-ME
1,6-Bis(maleimido)hexane 97%
BMH (1,6-Bis(maleimido)hexane)
Formula: C₁₄H₁₆N₂O₄
MW: 276.29 g/mol
Melting Pt: 144 °C
Storage Temperature: Ambient
MDL Number: MFCD00047122
CAS Number: 4856-87-5
EINECS: 225-449-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR