2-Amino-4-(2-furyl)thiazole 97%

Warning

H58851-06 H58851-14
AAH58851-14EA 1374.18 USD
AAH58851-14 AAH58851-06
2-Amino-4-(2-furyl)thiazole 97%
2-Amino-4-(2-furyl)thiazole
Formula: C₇H₆N₂OS
MW: 166.20 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01167261
CAS Number: 28989-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR