3-Aminoimidazo[1,2-a]pyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Imidazo[1,2-a]pyridin-3-amine

H58726-03 H58726-06
AAH58726-03EA 324.99 USD
AAH58726-03 AAH58726-06
3-Aminoimidazo[1,2-a]pyridine 97%
3-Aminoimidazo[1,2-a]pyridine
Formula: C₇H₇N₃
MW: 133.15 g/mol
Melting Pt: 114…126 °C
Storage Temperature: Ambient
MDL Number: MFCD06660421
CAS Number: 28036-33-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR