N-Cyclopropyl-4-nitrobenzenesulfonamide 97%

Supplier: Thermo Scientific Chemicals

Warning

H58673-ME H58673-MC
AAH58673-MEEA 289 USD
AAH58673-ME AAH58673-MC
N-Cyclopropyl-4-nitrobenzenesulfonamide 97%
N-Cyclopropyl-4-nitrobenzenesulphonamide
Formula: C₉H₁₀N₂O₄S
Storage Temperature: Ambient
MDL Number: MFCD00458256
CAS Number: 549476-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR