2-Amino-4-(4-bromophenyl)-5-methylthiazole 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-(4-Bromophenyl)-5-methylthiazol-2-amine

H58657-14 H58657-06
AAH58657-14EA 1144.41 USD
AAH58657-14 AAH58657-06
2-Amino-4-(4-bromophenyl)-5-methylthiazole 97%
2-Amino-4-(4-bromophenyl)-5-methylthiazole
Formula: C₁₀H₉BrN₂S
MW: 269.16 g/mol
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00170268
CAS Number: 65705-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR