4-(Trifluoromethoxy)thiobenzamide 97%

Warning

H58585-06 H58585-03
AAH58585-03EA 156.56 USD
AAH58585-03 AAH58585-06
4-(Trifluoromethoxy)thiobenzamide 97%
4-(Trifluoromethoxy)thiobenzamide
Formula: C₈H₆F₃NOS
Melting Pt: 124…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00068113
CAS Number: 149169-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR