3-Amino-2,5-dichloropyridine 97%

Danger

H58570-03 H58570-06 H58570-MD
AAH58570-03EA 97.42 USD
AAH58570-03 AAH58570-MD AAH58570-06
3-Amino-2,5-dichloropyridine 97%
3-Amino-2,5-dichloropyridine
Formula: C₅H₄Cl₂N₂
Melting Pt: 124…129 °C
Storage Temperature: Ambient
MDL Number: MFCD00204157
CAS Number: 78607-32-6
UN: 2811
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR